Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002493.4 | NP_002484.1 | 3 - 4 | 138 | |
GenBank mRNA | AF035840.1 | - | 3 - 4 | 138 | |
AF067167.1 | - | 3 - 4 | 138 | ||
AK289702.1 | - | 3 - 4 | 138 | ||
BC009801.1 | - | 3 - 4 | 138 | ||
BC016708.1 | - | 3 - 4 | 138 | ||
BC029247.1 | - | 3 - 4 | 138 | ||
BP208846.1 | - | 3 - 4 | 138 | ||
BU952486.1 | - | 1 - 2 | 424 | 138 | |
CR456920.1 | - | 3 - 4 | 138 |