Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022755.5 | NP_073592.1 | 9 - 10 | 1191 | 79 |
XM_017015041.1 | XP_016870530.1 | 9 - 10 | 1185 | 79 | |
GenBank mRNA | AF351202.1 | - | 9 - 10 | 997 | 79 |
AF520811.1 | - | 9 - 10 | 914 | 79 | |
AK023225.1 | - | 9 - 10 | 981 | 79 | |
BC026154.2 | - | 9 - 10 | 1193 | 79 |