Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042517.1 | NP_001035982.1 | 17 - 18 | 2293 | 161 |
NM_001258366.1 | NP_001245295.1 | 16 - 17 | 2260 | 161 | |
NM_001258367.1 | NP_001245296.1 | 15 - 16 | 2155 | 161 | |
NM_001258368.1 | NP_001245297.1 | 15 - 16 | 2083 | 161 | |
NM_001258369.1 | NP_001245298.1 | 17 - 18 | 2293 | 161 | |
NM_001258370.1 | NP_001245299.1 | 11 - 12 | 1464 | 161 | |
NM_030932.3 | NP_112194.2 | 11 - 12 | 1464 | 161 | |
XM_006719876.1 | XP_006719939.1 | 11 - 12 | 1444 | 161 | |
XM_011535258.2 | XP_011533560.1 | 17 - 18 | 2236 | 161 | |
GenBank mRNA | AB244756.1 | - | 16 - 17 | 2041 | 161 |
AB244757.1 | - | 15 - 16 | 1936 | 161 | |
AB244758.1 | - | 15 - 16 | 1864 | 161 | |
AK298509.1 | - | 17 - 18 | 2079 | 161 | |
AK307652.1 | - | 11 - 12 | 1444 | 161 | |
AK316474.1 | - | 17 - 18 | 2206 | 161 | |
AL137718.1 | - | 11 - 12 | 1436 | 161 | |
AY750055.1 | - | 17 - 18 | 2166 | 161 | |
AY818645.1 | - | 17 - 18 | 2074 | 161 | |
BC034952.1 | - | 11 - 12 | 1468 | 161 | |
BC048963.1 | - | 11 - 12 | 1420 | 161 | |
BC068504.1 | - | 11 - 12 | 1420 | 161 |