Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042517.1 | NP_001035982.1 | 8 - 9 | 1120 | 121 |
NM_001258366.1 | NP_001245295.1 | 7 - 8 | 1087 | 121 | |
NM_001258367.1 | NP_001245296.1 | 6 - 7 | 982 | 121 | |
NM_001258368.1 | NP_001245297.1 | 6 - 7 | 910 | 121 | |
NM_001258369.1 | NP_001245298.1 | 8 - 9 | 1120 | 121 | |
NM_001258370.1 | NP_001245299.1 | 2 - 3 | 291 | 121 | |
NM_030932.3 | NP_112194.2 | 2 - 3 | 291 | 121 | |
XM_006719876.1 | XP_006719939.1 | 2 - 3 | 271 | 121 | |
XM_011535258.2 | XP_011533560.1 | 8 - 9 | 1063 | 121 | |
XM_011535263.2 | XP_011533565.1 | 8 - 9 | 1063 | 121 | |
XM_011535265.2 | XP_011533567.1 | 8 - 9 | 1063 | 121 | |
XM_017020789.1 | XP_016876278.1 | 8 - 9 | 1063 | 121 | |
GenBank mRNA | AB244756.1 | - | 7 - 8 | 868 | 121 |
AB244758.1 | - | 6 - 7 | 691 | 121 | |
AK092024.1 | - | 9 - 10 | 1042 | 121 | |
AK298509.1 | - | 8 - 9 | 906 | 121 | |
AK307652.1 | - | 2 - 3 | 271 | 121 | |
AK316474.1 | - | 8 - 9 | 1033 | 121 | |
AL137718.1 | - | 2 - 3 | 263 | 121 | |
AY750055.1 | - | 8 - 9 | 993 | 121 | |
AY818645.1 | - | 8 - 9 | 901 | 121 | |
BC034952.1 | - | 2 - 3 | 295 | 121 | |
BC048963.1 | - | 2 - 3 | 247 | 121 | |
BC068504.1 | - | 2 - 3 | 247 | 121 | |
BX649186.1 | - | 8 - 9 | 1062 | 121 |