Assay Design
Gene Transcripts
- Gene Symbol
- SDCCAG8
- Entrez Gene ID
-
10806
- Gene Name
- serologically defined colon cancer antigen 8
- Gene Aliases
-
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
- Chromosome Location
-
Chr.1: 243255328 - 243500092 on Build GRCh38
View Full Product Details