Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152372.3 | NP_689585.3 | 11 - 12 | 1395 | 59 |
GenBank mRNA | AK125972.1 | - | 9 - 10 | 1437 | 59 |
AK128233.1 | - | 3 - 4 | 455 | 59 | |
BC067101.1 | - | 11 - 12 | 1405 | 59 | |
BX537911.1 | - | 10 - 11 | 1119 | 59 | |
BX647453.1 | - | 11 - 12 | 1356 | 59 | |
BX647455.1 | - | 11 - 12 | 1395 | 59 | |
BX647644.1 | - | 11 - 12 | 1356 | 59 |