Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152372.3 | NP_689585.3 | 12 - 13 | 1589 | 93 |
GenBank mRNA | AK125972.1 | - | 10 - 11 | 1631 | 93 |
AK128233.1 | - | 4 - 5 | 649 | 93 | |
BC067101.1 | - | 12 - 13 | 1599 | 93 | |
BX537911.1 | - | 11 - 12 | 1313 | 93 | |
BX647453.1 | - | 12 - 13 | 1550 | 93 | |
BX647455.1 | - | 12 - 13 | 1589 | 93 | |
BX647644.1 | - | 12 - 13 | 1550 | 93 |