Assay Design



Gene Transcripts

Gene Symbol
HP1BP3
Entrez Gene ID
50809
Gene Name
heterochromatin protein 1 binding protein 3
Gene Aliases
HP1-BP74, HP1BP74
Chromosome Location
Chr.1: 20742677 - 20787809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005245875.4 XP_005245932.1 1 - 2 613 140

View Full Product Details