Assay Design



Gene Transcripts

Gene Symbol
ANGEL2
Entrez Gene ID
90806
Gene Name
angel homolog 2
Gene Aliases
Ccr4d, KIAA0759L
Chromosome Location
Chr.1: 212992182 - 213015875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300753.1 NP_001287682.1 4 - 5 837 68
NM_001300755.1 NP_001287684.1 4 - 5 935 68
NM_001300757.1 NP_001287686.1 3 - 4 590 68
NM_001300758.1 NP_001287687.1 3 - 4 586 68
NM_144567.4 NP_653168.2 4 - 5 912 68
NR_125333.1 3 - 4 586 68
XM_005273344.1 XP_005273401.1 4 - 5 933 68
XM_005273345.1 XP_005273402.1 4 - 5 979 68
XM_005273346.2 XP_005273403.1 4 - 5 732 68
XM_005273347.3 XP_005273404.1 3 - 4 604 68
XM_017002774.1 XP_016858263.1 4 - 5 861 68
XM_017002775.1 XP_016858264.1 5 - 6 1160 68
XM_017002776.1 XP_016858265.1 5 - 6 1184 68
XM_017002777.1 XP_016858266.1 5 - 6 1230 68
XM_017002778.1 XP_016858267.1 5 - 6 973 68
GenBank mRNA AF510741.1 - 3 - 4 541 68
AK294479.1 - 4 - 5 711 68
AK295116.1 - 4 - 5 841 68
AK299208.1 - 4 - 5 905 68
AK299365.1 - 3 - 4 496 68
AK310946.1 - 2 - 3 78 68
AK311276.1 - 3 - 4 495 68
AL079275.1 - 4 - 5 870 68
BC047469.1 - 3 - 4 502 68

View Full Product Details