Assay Design



Gene Transcripts

Gene Symbol
TTC27
Entrez Gene ID
55622
Gene Name
tetratricopeptide repeat domain 27
Gene Aliases
-
Chromosome Location
Chr.2: 32628020 - 32821051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193509.1 NP_001180438.1 7 - 8 964 96
NM_017735.4 NP_060205.3 7 - 8 1211 96
XM_005264416.1 XP_005264473.1 7 - 8 1198 96
XM_011532958.1 XP_011531260.1 7 - 8 1198 96
GenBank mRNA AK000279.1 - 4 - 5 419 96
AK000665.1 - 2 - 3 203 96
AK027570.1 - 7 - 8 1120 96
AK225295.1 - 4 - 5 419 96
AK299196.1 - 7 - 8 953 96
BC001248.1 - 6 - 7 744 96
BC063791.1 - 7 - 8 1169 96

View Full Product Details