Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098579.2 | NP_001092049.2 | 12 - 13 | 1798 | 76 |
NM_001100163.2 | NP_001093633.1 | 13 - 14 | 2076 | 76 | |
NM_001100167.2 | NP_001093637.1 | 13 - 14 | 1793 | 76 | |
NM_001206880.1 | NP_001193809.1 | 12 - 13 | 1511 | 76 | |
NM_001206881.1 | NP_001193810.1 | 11 - 12 | 1918 | 76 | |
NM_130385.3 | NP_569056.4 | 13 - 14 | 2129 | 76 | |
GenBank mRNA | AF081249.1 | - | 11 - 12 | 1814 | 76 |
AF081250.1 | - | 11 - 12 | 1542 | 76 | |
AK127209.1 | - | 13 - 14 | 1771 | 76 | |
AK296336.1 | - | 13 - 14 | 1893 | 76 | |
AK300358.1 | - | 12 - 13 | 1783 | 76 | |
AK301296.1 | - | 12 - 13 | 1122 | 76 | |
AK304702.1 | - | 12 - 13 | 1145 | 76 | |
BC117127.1 | - | 13 - 14 | 1666 | 76 |