Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098579.2 | NP_001092049.2 | 13 - 14 | 1934 | 63 |
NM_001100163.2 | NP_001093633.1 | 14 - 15 | 2212 | 63 | |
NM_001100167.2 | NP_001093637.1 | 14 - 15 | 1929 | 63 | |
NM_001206880.1 | NP_001193809.1 | 13 - 14 | 1647 | 63 | |
NM_001206881.1 | NP_001193810.1 | 12 - 13 | 2054 | 63 | |
NM_130385.3 | NP_569056.4 | 14 - 15 | 2265 | 63 | |
GenBank mRNA | AF081249.1 | - | 12 - 13 | 1950 | 63 |
AF081250.1 | - | 12 - 13 | 1678 | 63 | |
AK127209.1 | - | 14 - 15 | 1907 | 63 | |
AK300358.1 | - | 13 - 14 | 1919 | 63 | |
AK301296.1 | - | 13 - 14 | 1258 | 63 | |
AK304702.1 | - | 13 - 14 | 1281 | 63 | |
BC117127.1 | - | 14 - 15 | 1802 | 63 |