Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098579.2 | NP_001092049.2 | 18 - 19 | 2414 | 88 |
NM_001100163.2 | NP_001093633.1 | 19 - 20 | 2692 | 88 | |
NM_001100167.2 | NP_001093637.1 | 19 - 20 | 2409 | 88 | |
NM_001206880.1 | NP_001193809.1 | 18 - 19 | 2127 | 88 | |
NM_001206881.1 | NP_001193810.1 | 17 - 18 | 2534 | 88 | |
NM_130385.3 | NP_569056.4 | 19 - 20 | 2745 | 88 | |
GenBank mRNA | AF081249.1 | - | 17 - 18 | 2430 | 88 |
AF081250.1 | - | 17 - 18 | 2158 | 88 | |
AK127209.1 | - | 19 - 20 | 2387 | 88 | |
AK296336.1 | - | 19 - 20 | 2509 | 88 | |
AK300358.1 | - | 18 - 19 | 2399 | 88 | |
AK304702.1 | - | 18 - 19 | 1761 | 88 | |
BC117127.1 | - | 19 - 20 | 2282 | 88 |