Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003019.4 | NP_003010.4 | 7 - 8 | 793 | 62 |
XM_011540087.1 | XP_011538389.1 | 7 - 8 | 772 | 62 | |
XM_011540088.2 | XP_011538390.1 | 6 - 7 | 744 | 62 | |
GenBank mRNA | BC022318.1 | - | 7 - 8 | 794 | 62 |
BM985387.1 | - | 1 - 2 | 507 | 62 | |
CR541948.1 | - | 6 - 7 | 750 | 62 | |
X65018.1 | - | 7 - 8 | 921 | 62 |