Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003594.3 | NP_003585.3 | 3 - 4 | 236 | 67 |
XM_005271277.3 | XP_005271334.1 | 3 - 4 | 275 | 67 | |
XM_011542303.2 | XP_011540605.1 | 3 - 4 | 275 | 67 | |
XM_017002549.1 | XP_016858038.1 | 3 - 4 | 275 | 67 | |
XM_017002550.1 | XP_016858039.1 | 3 - 4 | 275 | 67 | |
XM_017002551.1 | XP_016858040.1 | 3 - 4 | 275 | 67 | |
XM_017002552.1 | XP_016858041.1 | 3 - 4 | 275 | 67 | |
XM_017002553.1 | XP_016858042.1 | 3 - 4 | 275 | 67 | |
XM_017002554.1 | XP_016858043.1 | 3 - 4 | 275 | 67 | |
XM_017002555.1 | XP_016858044.1 | 3 - 4 | 275 | 67 | |
GenBank mRNA | AF073771.1 | - | 3 - 4 | 236 | 67 |
AK291017.1 | - | 3 - 4 | 275 | 67 | |
BC030058.1 | - | 3 - 4 | 220 | 67 |