Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020650.2 | NP_065701.2 | 4 - 5 | 1068 | 71 |
XM_005259089.3 | XP_005259146.1 | 5 - 6 | 1163 | 71 | |
XM_011527143.1 | XP_011525445.1 | 5 - 6 | 1257 | 71 | |
XM_017027023.1 | XP_016882512.1 | 4 - 5 | 1342 | 71 | |
GenBank mRNA | AF183423.1 | - | 3 - 4 | 653 | 71 |
AY195859.1 | - | 4 - 5 | 1068 | 71 | |
AY358423.1 | - | 4 - 5 | 655 | 71 | |
BC013436.1 | - | 4 - 5 | 651 | 71 |