Assay Design



Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024867.3 NP_079143.3 34 - 35 5115 63
XM_005248376.3 XP_005248433.1 34 - 35 5220 63
XM_005248377.3 XP_005248434.1 34 - 35 5205 63
XM_011514135.2 XP_011512437.1 39 - 40 6047 63
XM_011514136.2 XP_011512438.1 39 - 40 6046 63
XM_011514137.2 XP_011512439.1 39 - 40 6031 63
XM_011514138.2 XP_011512440.1 39 - 40 6023 63
XM_011514139.2 XP_011512441.1 38 - 39 5971 63
XM_011514140.2 XP_011512442.1 38 - 39 5781 63
XM_011514141.2 XP_011512443.1 40 - 41 6245 63
XM_011514142.1 XP_011512444.1 34 - 35 5196 63
XM_017009880.1 XP_016865369.1 38 - 39 5863 63
XM_017009881.1 XP_016865370.1 39 - 40 6901 63
GenBank mRNA AB051557.1 - 24 - 25 3456 63
AK026817.1 - 10 - 11 1619 63
BX648691.1 - 12 - 13 2623 63

View Full Product Details