Assay Design



Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514135.2 XP_011512437.1 33 - 34 5173 111
XM_011514136.2 XP_011512438.1 33 - 34 5172 111
XM_011514137.2 XP_011512439.1 33 - 34 5157 111
XM_011514138.2 XP_011512440.1 33 - 34 5149 111
XM_011514139.2 XP_011512441.1 32 - 33 5097 111
XM_011514140.2 XP_011512442.1 32 - 33 4907 111
XM_011514141.2 XP_011512443.1 34 - 35 5371 111
XM_017009880.1 XP_016865369.1 32 - 33 4989 111
XM_017009881.1 XP_016865370.1 33 - 34 6027 111
GenBank mRNA AK026817.1 - 4 - 5 745 111
BX648691.1 - 6 - 7 1749 111

View Full Product Details