Assay Design
Gene Transcripts
- Gene Symbol
- FIZ1
- Entrez Gene ID
-
84922
- Gene Name
- FLT3 interacting zinc finger 1
- Gene Aliases
-
ZNF798
- Chromosome Location
-
Chr.19: 55591371 - 55599703 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_032836.2 |
NP_116225.2 |
1 - 2 |
48 |
168 |
GenBank mRNA |
AK027674.1 |
- |
1 - 2 |
48 |
168 |
View Full Product Details