Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015132.4 | NP_055947.1 | 9 - 10 | 1052 | 133 |
XM_005249672.1 | XP_005249729.1 | 9 - 10 | 1062 | 133 | |
XM_005249673.4 | XP_005249730.1 | 9 - 10 | 1062 | 133 | |
XM_011515229.1 | XP_011513531.1 | 9 - 10 | 1062 | 133 | |
XM_011515230.2 | XP_011513532.1 | 10 - 11 | 1109 | 133 | |
XM_011515231.2 | XP_011513533.1 | 11 - 12 | 1168 | 133 | |
XM_011515233.2 | XP_011513535.1 | 10 - 11 | 1110 | 133 | |
XM_017011893.1 | XP_016867382.1 | 9 - 10 | 1062 | 133 | |
XM_017011894.1 | XP_016867383.1 | 10 - 11 | 1509 | 133 | |
XM_017011895.1 | XP_016867384.1 | 10 - 11 | 1114 | 133 | |
XM_017011896.1 | XP_016867385.1 | 10 - 11 | 3022 | 133 | |
XM_017011897.1 | XP_016867386.1 | 9 - 10 | 983 | 133 | |
GenBank mRNA | AB018256.1 | - | 9 - 10 | 1012 | 133 |
AF420470.1 | - | 9 - 10 | 1020 | 133 | |
AK023740.1 | - | 4 - 5 | 297 | 133 | |
AK294582.1 | - | 11 - 12 | 1007 | 133 | |
AK315135.1 | - | 9 - 10 | 1038 | 133 | |
BC045667.1 | - | 9 - 10 | 1022 | 133 |