Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016616.4 | NP_057700.3 | 11 - 12 | 1189 | 94 |
GenBank mRNA | AF202051.2 | - | 10 - 11 | 918 | 94 |
AF305596.1 | - | 10 - 11 | 887 | 94 | |
AK313599.1 | - | 10 - 11 | 929 | 94 | |
BC036816.1 | - | 11 - 12 | 1189 | 94 | |
GQ472221.1 | - | 11 - 12 | 1189 | 94 |