Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016616.4 | NP_057700.3 | 9 - 10 | 907 | 103 |
GenBank mRNA | AF202051.2 | - | 8 - 9 | 636 | 103 |
AF305596.1 | - | 8 - 9 | 605 | 103 | |
AK313599.1 | - | 8 - 9 | 647 | 103 | |
BC036816.1 | - | 9 - 10 | 907 | 103 | |
GQ472221.1 | - | 9 - 10 | 907 | 103 |