Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002133.2 | NP_002124.1 | 4 - 5 | 842 | 130 |
GenBank mRNA | AK313120.1 | - | 4 - 5 | 812 | 130 |
BC001491.2 | - | 4 - 5 | 801 | 130 | |
BT019785.1 | - | 3 - 4 | 735 | 130 | |
CR456505.1 | - | 4 - 5 | 850 | 130 | |
CR541945.1 | - | 4 - 5 | 735 | 130 | |
CR541968.1 | - | 4 - 5 | 735 | 130 | |
JF323038.1 | - | 4 - 5 | 739 | 130 | |
X06985.1 | - | 4 - 5 | 815 | 130 |