Assay Design
Gene Transcripts
- Gene Symbol
- FREM1
- Entrez Gene ID
-
158326
- Gene Name
- FRAS1 related extracellular matrix 1
- Gene Aliases
-
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
- Chromosome Location
-
Chr.9: 14734666 - 14911653 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK058190.1 |
- |
1 - 2 |
94 |
111 |
View Full Product Details