Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001851.2 | NP_001001851.1 | 1 - 2 | 194 | 106 |
NM_030569.6 | NP_085046.5 | 1 - 2 | 194 | 106 | |
XM_011519713.2 | XP_011518015.1 | 1 - 2 | 194 | 106 | |
XM_011519714.2 | XP_011518016.1 | 1 - 2 | 194 | 106 | |
GenBank mRNA | AK027375.1 | - | 1 - 2 | 131 | 106 |
AK075381.1 | - | 1 - 2 | 167 | 106 | |
AY238437.1 | - | 1 - 2 | 90 | 106 | |
AY358426.1 | - | 1 - 2 | 159 | 106 | |
BC136439.1 | - | 1 - 2 | 167 | 106 | |
DA246953.1 | - | 1 - 2 | 194 | 106 |