Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000022.3 | NP_000013.2 | 2 - 3 | 247 | 76 |
NM_001322050.1 | NP_001308979.1 | 2 - 3 | 247 | 76 | |
NM_001322051.1 | NP_001308980.1 | 2 - 3 | 247 | 76 | |
NR_136160.1 | 2 - 3 | 247 | 76 | ||
GenBank mRNA | AK223397.1 | - | 2 - 3 | 197 | 76 |
BC007678.2 | - | 2 - 3 | 208 | 76 | |
BC040226.1 | - | 2 - 3 | 214 | 76 | |
BI601734.1 | - | 2 - 3 | 236 | 76 | |
CD014053.1 | - | 2 - 3 | 180 | 76 | |
CD014056.1 | - | 2 - 3 | 180 | 76 | |
HY102642.1 | - | 2 - 3 | 248 | 76 | |
KJ534760.1 | - | 2 - 3 | 96 | 76 | |
KU177892.1 | - | 2 - 3 | 96 | 76 | |
KU177893.1 | - | 2 - 3 | 96 | 76 | |
KU177894.1 | - | 2 - 3 | 96 | 76 | |
KU177895.1 | - | 2 - 3 | 96 | 76 | |
X02994.1 | - | 2 - 3 | 191 | 76 |