Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145353.1 | NP_001138825.1 | 5 - 6 | 772 | 132 |
NM_172373.3 | NP_758961.1 | 6 - 7 | 989 | 132 | |
XM_005266276.1 | XP_005266333.1 | 6 - 7 | 878 | 132 | |
XM_005266277.3 | XP_005266334.1 | 6 - 7 | 1421 | 132 | |
XM_011534950.1 | XP_011533252.1 | 8 - 9 | 1227 | 132 | |
XM_011534951.2 | XP_011533253.1 | 7 - 8 | 1540 | 132 | |
XM_017020409.1 | XP_016875898.1 | 8 - 9 | 1207 | 132 | |
XM_017020410.1 | XP_016875899.1 | 7 - 8 | 1144 | 132 | |
XM_017020411.1 | XP_016875900.1 | 7 - 8 | 1090 | 132 | |
XM_017020412.1 | XP_016875901.1 | 7 - 8 | 1218 | 132 | |
XM_017020413.1 | XP_016875902.1 | 7 - 8 | 1272 | 132 | |
GenBank mRNA | AK304289.1 | - | 5 - 6 | 132 | |
BC030507.2 | - | 5 - 6 | 844 | 132 | |
BX640798.1 | - | 3 - 4 | 270 | 132 | |
M82882.1 | - | 6 - 7 | 931 | 132 |