Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145353.1 | NP_001138825.1 | 6 - 7 | 960 | 111 |
NM_172373.3 | NP_758961.1 | 7 - 8 | 1177 | 111 | |
XM_005266276.1 | XP_005266333.1 | 7 - 8 | 1066 | 111 | |
XM_005266277.3 | XP_005266334.1 | 7 - 8 | 1609 | 111 | |
XM_011534950.1 | XP_011533252.1 | 9 - 10 | 1415 | 111 | |
XM_011534951.2 | XP_011533253.1 | 8 - 9 | 1728 | 111 | |
XM_017020409.1 | XP_016875898.1 | 9 - 10 | 1395 | 111 | |
XM_017020410.1 | XP_016875899.1 | 8 - 9 | 1332 | 111 | |
XM_017020411.1 | XP_016875900.1 | 8 - 9 | 1278 | 111 | |
XM_017020412.1 | XP_016875901.1 | 8 - 9 | 1406 | 111 | |
XM_017020413.1 | XP_016875902.1 | 8 - 9 | 1460 | 111 | |
GenBank mRNA | AK304289.1 | - | 6 - 7 | 903 | 111 |
BC030507.2 | - | 6 - 7 | 1032 | 111 | |
BX640798.1 | - | 4 - 5 | 458 | 111 | |
M82882.1 | - | 7 - 8 | 1119 | 111 |