Assay Design



Gene Transcripts

Gene Symbol
MAGI2
Entrez Gene ID
9863
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 2
Gene Aliases
ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM
Chromosome Location
Chr.7: 78017055 - 79453805 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301128.1 NP_001288057.1 15 - 16 3057 75
NM_012301.3 NP_036433.2 16 - 17 3099 75
XM_011516718.1 XP_011515020.1 16 - 17 3330 75
XM_011516719.2 XP_011515021.1 16 - 17 6747 75
XM_011516720.2 XP_011515022.1 16 - 17 11737 75
XM_011516726.2 XP_011515028.1 13 - 14 2403 75
XM_011516728.1 XP_011515030.1 12 - 13 1939 75
XM_017012840.1 XP_016868329.1 17 - 18 3459 75
XM_017012841.1 XP_016868330.1 17 - 18 3456 75
XM_017012842.1 XP_016868331.1 17 - 18 3453 75
XM_017012843.1 XP_016868332.1 16 - 17 3417 75
XM_017012844.1 XP_016868333.1 17 - 18 3459 75
XM_017012845.1 XP_016868334.1 16 - 17 3324 75
XM_017012846.1 XP_016868335.1 15 - 16 3288 75
XM_017012847.1 XP_016868336.1 17 - 18 9696 75
XM_017012848.1 XP_016868337.1 15 - 16 6615 75
XM_017012849.1 XP_016868338.1 15 - 16 6609 75
XM_017012850.1 XP_016868339.1 17 - 18 3459 75
XM_017012851.1 XP_016868340.1 17 - 18 3459 75
XM_017012852.1 XP_016868341.1 17 - 18 3459 75
GenBank mRNA AB014605.2 - 15 - 16 3038 75
AK297353.1 - 16 - 17 3084 75
AK309012.1 - 11 - 12 1893 75
BC150277.1 - 15 - 16 3038 75

View Full Product Details