Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011516718.1 | XP_011515020.1 | 21 - 22 | 4191 | 88 |
XM_011516719.2 | XP_011515021.1 | 21 - 22 | 7608 | 88 | |
XM_011516720.2 | XP_011515022.1 | 21 - 22 | 12598 | 88 | |
XM_011516726.2 | XP_011515028.1 | 18 - 19 | 3264 | 88 | |
XM_011516728.1 | XP_011515030.1 | 17 - 18 | 2800 | 88 | |
XM_017012840.1 | XP_016868329.1 | 22 - 23 | 4320 | 88 | |
XM_017012841.1 | XP_016868330.1 | 22 - 23 | 4317 | 88 | |
XM_017012842.1 | XP_016868331.1 | 22 - 23 | 4314 | 88 | |
XM_017012843.1 | XP_016868332.1 | 21 - 22 | 4278 | 88 | |
XM_017012845.1 | XP_016868334.1 | 21 - 22 | 4185 | 88 | |
XM_017012846.1 | XP_016868335.1 | 20 - 21 | 4149 | 88 | |
XM_017012847.1 | XP_016868336.1 | 22 - 23 | 10557 | 88 | |
XM_017012848.1 | XP_016868337.1 | 20 - 21 | 7476 | 88 | |
XM_017012849.1 | XP_016868338.1 | 20 - 21 | 7470 | 88 | |
XM_017012850.1 | XP_016868339.1 | 22 - 23 | 4320 | 88 |