Assay Design
Gene Transcripts
- Gene Symbol
- MAGI2
- Entrez Gene ID
-
9863
- Gene Name
- membrane associated guanylate kinase, WW and PDZ domain containing 2
- Gene Aliases
-
ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM
- Chromosome Location
-
Chr.7: 78017055 - 79453805 on Build GRCh38
View Full Product Details