Assay Design



Gene Transcripts

Gene Symbol
ERLIN1
Entrez Gene ID
10613
Gene Name
ER lipid raft associated 1
Gene Aliases
C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62
Chromosome Location
Chr.10: 100150090 - 100186057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100626.1 NP_001094096.1 9 - 10 759 130
NM_006459.3 NP_006450.2 8 - 9 1003 130
XM_005269442.3 XP_005269499.1 9 - 10 789 130
XM_017015541.1 XP_016871030.1 9 - 10 773 130
GenBank mRNA AF064093.1 - 9 - 10 735 130
AK056574.1 - 8 - 9 3360 130
AK127017.1 - 2 - 3 2963 130
AK222480.1 - 9 - 10 731 130
AK298427.1 - 8 - 9 649 130
AK315580.1 - 9 - 10 731 130
BC031791.1 - 8 - 9 672 130

View Full Product Details