Assay Design



Gene Transcripts

Gene Symbol
CAMSAP1
Entrez Gene ID
157922
Gene Name
calmodulin regulated spectrin associated protein 1
Gene Aliases
-
Chromosome Location
Chr.9: 135808487 - 135907551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015447.3 NP_056262.3 9 - 10 1317 68
XM_005263396.3 XP_005263453.1 10 - 11 1742 68
XM_005263397.1 XP_005263454.1 7 - 8 843 68
XM_011518272.1 XP_011516574.1 9 - 10 1011 68
XM_011518273.2 XP_011516575.1 8 - 9 842 68
XM_011518274.1 XP_011516576.1 7 - 8 872 68
XM_017014301.1 XP_016869790.1 9 - 10 1598 68
XM_017014302.1 XP_016869791.1 9 - 10 1522 68
GenBank mRNA AJ519841.1 - 7 - 8 842 68
BC130580.1 - 9 - 10 976 68
BC144082.1 - 9 - 10 976 68

View Full Product Details