Assay Design



Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 7 - 8 971 64
NM_001168408.1 NP_001161880.1 7 - 8 971 64
NM_001168409.1 NP_001161881.1 7 - 8 756 64
NM_001168410.1 NP_001161882.1 7 - 8 1043 64
NM_014989.5 NP_055804.2 12 - 13 2558 64
XM_005248688.3 XP_005248745.1 12 - 13 2494 64
XM_011535604.2 XP_011533906.2 12 - 13 2494 64
XM_011535611.2 XP_011533913.2 12 - 13 2494 64
XM_011535615.2 XP_011533917.2 12 - 13 2494 64
XM_017010516.1 XP_016866005.1 12 - 13 2494 64
XM_017010517.1 XP_016866006.1 12 - 13 2494 64
XM_017010518.1 XP_016866007.1 12 - 13 2494 64
XM_017010519.1 XP_016866008.1 12 - 13 2494 64
XM_017010520.1 XP_016866009.1 12 - 13 2494 64
XM_017010521.1 XP_016866010.1 12 - 13 2494 64
XM_017010522.1 XP_016866011.1 12 - 13 2494 64
XM_017010523.1 XP_016866012.1 12 - 13 2494 64
XM_017010524.1 XP_016866013.1 12 - 13 2494 64
XM_017010525.1 XP_016866014.1 12 - 13 2494 64
XM_017010526.1 XP_016866015.1 12 - 13 2494 64
XM_017010527.1 XP_016866016.1 12 - 13 2494 64
XM_017010528.1 XP_016866017.1 12 - 13 2494 64
XM_017010529.1 XP_016866018.1 12 - 13 2494 64
XM_017010530.1 XP_016866019.1 12 - 13 2494 64
XM_017010531.1 XP_016866020.1 12 - 13 2494 64
XM_017010532.1 XP_016866021.1 12 - 13 2494 64
XM_017010533.1 XP_016866022.1 12 - 13 2494 64
XM_017010534.1 XP_016866023.1 7 - 8 1067 64
XM_017010535.1 XP_016866024.1 7 - 8 1067 64
XM_017010536.1 XP_016866025.1 7 - 8 1064 64
XM_017010537.1 XP_016866026.1 7 - 8 1064 64
XM_017010538.1 XP_016866027.1 7 - 8 1172 64
XM_017010539.1 XP_016866028.1 7 - 8 1059 64
XM_017010540.1 XP_016866029.1 7 - 8 1058 64
XM_017010541.1 XP_016866030.1 7 - 8 1058 64
XM_017010542.1 XP_016866031.1 7 - 8 1058 64
XM_017010543.1 XP_016866032.1 7 - 8 977 64
XM_017010544.1 XP_016866033.1 7 - 8 1060 64
XM_017010545.1 XP_016866034.1 7 - 8 1057 64
XM_017010546.1 XP_016866035.1 7 - 8 1170 64
XM_017010547.1 XP_016866036.1 7 - 8 1170 64
XM_017010548.1 XP_016866037.1 7 - 8 1055 64
XM_017010549.1 XP_016866038.1 7 - 8 1055 64
XM_017010550.1 XP_016866039.1 7 - 8 1438 64
XM_017010551.1 XP_016866040.1 7 - 8 1438 64
XM_017010552.1 XP_016866041.1 7 - 8 1295 64
XM_017010553.1 XP_016866042.1 7 - 8 1295 64
XM_017010554.1 XP_016866043.1 7 - 8 1291 64
XM_017010555.1 XP_016866044.1 7 - 8 1291 64
GenBank mRNA AB002338.1 - 13 - 14 2281 64
AB045726.1 - 13 - 14 2524 64
AB051866.1 - 13 - 14 2524 64
AK293828.1 - 7 - 8 646 64
AK294868.1 - 7 - 8 971 64
AK295001.1 - 7 - 8 889 64
AK295177.1 - 7 - 8 957 64
AK296303.1 - 7 - 8 812 64
AK308658.1 - 7 - 8 1043 64
AK309185.1 - 7 - 8 756 64
AK311105.1 - 7 - 8 918 64
AK316108.1 - 7 - 8 769 64
AY190519.1 - 12 - 13 2237 64
BC151853.1 - 13 - 14 2305 64
BC152435.1 - 13 - 14 2281 64

View Full Product Details