Assay Design



Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 9 - 10 1273 72
NM_001168408.1 NP_001161880.1 9 - 10 1273 72
NM_001168409.1 NP_001161881.1 9 - 10 1058 72
NM_001168410.1 NP_001161882.1 9 - 10 1345 72
NM_014989.5 NP_055804.2 14 - 15 2860 72
XM_005248688.3 XP_005248745.1 14 - 15 2796 72
XM_011535604.2 XP_011533906.2 14 - 15 2796 72
XM_011535611.2 XP_011533913.2 14 - 15 2796 72
XM_011535615.2 XP_011533917.2 14 - 15 2796 72
XM_017010516.1 XP_016866005.1 14 - 15 2796 72
XM_017010517.1 XP_016866006.1 14 - 15 2796 72
XM_017010518.1 XP_016866007.1 14 - 15 2796 72
XM_017010519.1 XP_016866008.1 14 - 15 2796 72
XM_017010520.1 XP_016866009.1 14 - 15 2796 72
XM_017010521.1 XP_016866010.1 14 - 15 2796 72
XM_017010522.1 XP_016866011.1 14 - 15 2796 72
XM_017010523.1 XP_016866012.1 14 - 15 2796 72
XM_017010524.1 XP_016866013.1 14 - 15 2796 72
XM_017010525.1 XP_016866014.1 14 - 15 2796 72
XM_017010526.1 XP_016866015.1 14 - 15 2796 72
XM_017010527.1 XP_016866016.1 14 - 15 2796 72
XM_017010528.1 XP_016866017.1 14 - 15 2796 72
XM_017010529.1 XP_016866018.1 14 - 15 2796 72
XM_017010530.1 XP_016866019.1 14 - 15 2796 72
XM_017010531.1 XP_016866020.1 14 - 15 2796 72
XM_017010532.1 XP_016866021.1 14 - 15 2796 72
XM_017010533.1 XP_016866022.1 14 - 15 2796 72
XM_017010534.1 XP_016866023.1 9 - 10 1369 72
XM_017010535.1 XP_016866024.1 9 - 10 1369 72
XM_017010536.1 XP_016866025.1 9 - 10 1366 72
XM_017010537.1 XP_016866026.1 9 - 10 1366 72
XM_017010538.1 XP_016866027.1 9 - 10 1474 72
XM_017010539.1 XP_016866028.1 9 - 10 1361 72
XM_017010540.1 XP_016866029.1 9 - 10 1360 72
XM_017010541.1 XP_016866030.1 9 - 10 1360 72
XM_017010542.1 XP_016866031.1 9 - 10 1360 72
XM_017010543.1 XP_016866032.1 9 - 10 1279 72
XM_017010544.1 XP_016866033.1 9 - 10 1362 72
XM_017010545.1 XP_016866034.1 9 - 10 1359 72
XM_017010546.1 XP_016866035.1 9 - 10 1472 72
XM_017010547.1 XP_016866036.1 9 - 10 1472 72
XM_017010548.1 XP_016866037.1 9 - 10 1357 72
XM_017010549.1 XP_016866038.1 9 - 10 1357 72
XM_017010550.1 XP_016866039.1 9 - 10 1740 72
XM_017010551.1 XP_016866040.1 9 - 10 1740 72
XM_017010552.1 XP_016866041.1 9 - 10 1597 72
XM_017010553.1 XP_016866042.1 9 - 10 1597 72
XM_017010554.1 XP_016866043.1 9 - 10 1593 72
XM_017010555.1 XP_016866044.1 9 - 10 1593 72
GenBank mRNA AB002338.1 - 15 - 16 2583 72
AB045726.1 - 15 - 16 2826 72
AB051866.1 - 15 - 16 2826 72
AF263305.1 - 2 - 3 214 72
AF263306.1 - 2 - 3 214 72
AF263307.1 - 2 - 3 214 72
AF263308.1 - 2 - 3 214 72
AF263309.1 - 2 - 3 214 72
AF263310.1 - 2 - 3 214 72
AK293828.1 - 9 - 10 948 72
AK294868.1 - 9 - 10 1273 72
AK295001.1 - 9 - 10 1191 72
AK295177.1 - 9 - 10 1259 72
AK296303.1 - 9 - 10 1114 72
AK308658.1 - 9 - 10 1345 72
AK309185.1 - 9 - 10 1058 72
AK316108.1 - 9 - 10 1071 72
AY190519.1 - 14 - 15 2539 72
BC151853.1 - 15 - 16 2607 72
BC152435.1 - 15 - 16 2583 72

View Full Product Details