Assay Design



Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 13 - 14 1787 104
NM_001168408.1 NP_001161880.1 13 - 14 1790 104
NM_001168409.1 NP_001161881.1 13 - 14 1575 104
NM_001168410.1 NP_001161882.1 13 - 14 1862 104
NM_014989.5 NP_055804.2 18 - 19 3377 104
XM_005248688.3 XP_005248745.1 18 - 19 3313 104
XM_011535604.2 XP_011533906.2 18 - 19 3310 104
XM_011535615.2 XP_011533917.2 18 - 19 3310 104
XM_017010516.1 XP_016866005.1 18 - 19 3313 104
XM_017010517.1 XP_016866006.1 18 - 19 3313 104
XM_017010518.1 XP_016866007.1 18 - 19 3310 104
XM_017010519.1 XP_016866008.1 18 - 19 3313 104
XM_017010520.1 XP_016866009.1 18 - 19 3313 104
XM_017010521.1 XP_016866010.1 18 - 19 3313 104
XM_017010522.1 XP_016866011.1 18 - 19 3313 104
XM_017010523.1 XP_016866012.1 18 - 19 3313 104
XM_017010524.1 XP_016866013.1 18 - 19 3310 104
XM_017010525.1 XP_016866014.1 18 - 19 3313 104
XM_017010527.1 XP_016866016.1 18 - 19 3310 104
XM_017010530.1 XP_016866019.1 18 - 19 3313 104
XM_017010531.1 XP_016866020.1 18 - 19 3310 104
XM_017010532.1 XP_016866021.1 18 - 19 3313 104
XM_017010533.1 XP_016866022.1 18 - 19 3313 104
XM_017010534.1 XP_016866023.1 13 - 14 1886 104
XM_017010535.1 XP_016866024.1 13 - 14 1883 104
XM_017010536.1 XP_016866025.1 13 - 14 1883 104
XM_017010537.1 XP_016866026.1 13 - 14 1883 104
XM_017010538.1 XP_016866027.1 13 - 14 1991 104
XM_017010539.1 XP_016866028.1 13 - 14 1878 104
XM_017010540.1 XP_016866029.1 13 - 14 1877 104
XM_017010541.1 XP_016866030.1 13 - 14 1874 104
XM_017010542.1 XP_016866031.1 13 - 14 1877 104
XM_017010543.1 XP_016866032.1 13 - 14 1793 104
XM_017010545.1 XP_016866034.1 13 - 14 1873 104
XM_017010546.1 XP_016866035.1 13 - 14 1989 104
XM_017010547.1 XP_016866036.1 13 - 14 1986 104
XM_017010550.1 XP_016866039.1 13 - 14 2257 104
XM_017010551.1 XP_016866040.1 13 - 14 2254 104
XM_017010552.1 XP_016866041.1 13 - 14 2114 104
XM_017010553.1 XP_016866042.1 13 - 14 2111 104
XM_017010554.1 XP_016866043.1 13 - 14 2107 104
XM_017010555.1 XP_016866044.1 13 - 14 2107 104
XM_017010557.1 XP_016866046.1 1 - 2 113 104
GenBank mRNA AB002338.1 - 19 - 20 3100 104
AB045726.1 - 19 - 20 3343 104
AB051866.1 - 19 - 20 3343 104
AF263305.1 - 6 - 7 731 104
AF263306.1 - 6 - 7 731 104
AF263307.1 - 6 - 7 728 104
AF263308.1 - 6 - 7 731 104
AF263309.1 - 6 - 7 728 104
AK293828.1 - 13 - 14 1465 104
AK294868.1 - 13 - 14 1787 104
AK295001.1 - 13 - 14 1708 104
AK296303.1 - 13 - 14 1631 104
AK302571.1 - 3 - 4 328 104
AK308658.1 - 13 - 14 1862 104
AK309185.1 - 13 - 14 1575 104
AK316108.1 - 13 - 14 1588 104
AY190519.1 - 18 - 19 3056 104
BC151853.1 - 19 - 20 3124 104
BC152435.1 - 19 - 20 3100 104

View Full Product Details