Assay Design



Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 19 - 20 2403 115
NM_014989.5 NP_055804.2 28 - 29 4452 115
XM_005248688.3 XP_005248745.1 23 - 24 3857 115
XM_011535604.2 XP_011533906.2 24 - 25 3926 115
XM_011535611.2 XP_011533913.2 21 - 22 3701 115
XM_017010516.1 XP_016866005.1 26 - 27 4175 115
XM_017010517.1 XP_016866006.1 26 - 27 4082 115
XM_017010518.1 XP_016866007.1 26 - 27 4079 115
XM_017010519.1 XP_016866008.1 24 - 25 4022 115
XM_017010520.1 XP_016866009.1 25 - 26 4010 115
XM_017010521.1 XP_016866010.1 25 - 26 3992 115
XM_017010522.1 XP_016866011.1 24 - 25 3929 115
XM_017010524.1 XP_016866013.1 24 - 25 3884 115
XM_017010525.1 XP_016866014.1 24 - 25 3869 115
XM_017010526.1 XP_016866015.1 23 - 24 3857 115
XM_017010527.1 XP_016866016.1 23 - 24 3854 115
XM_017010528.1 XP_016866017.1 22 - 23 3731 115
XM_017010529.1 XP_016866018.1 21 - 22 3704 115
XM_017010534.1 XP_016866023.1 20 - 21 2685 115
XM_017010535.1 XP_016866024.1 20 - 21 2682 115
XM_017010536.1 XP_016866025.1 19 - 20 2592 115
XM_017010537.1 XP_016866026.1 20 - 21 2580 115
XM_017010539.1 XP_016866028.1 19 - 20 2494 115
XM_017010540.1 XP_016866029.1 19 - 20 2451 115
XM_017010541.1 XP_016866030.1 19 - 20 2448 115
XM_017010542.1 XP_016866031.1 18 - 19 2421 115
XM_017010543.1 XP_016866032.1 18 - 19 2337 115
XM_017010544.1 XP_016866033.1 18 - 19 2420 115
XM_017010545.1 XP_016866034.1 18 - 19 2399 115
XM_017010548.1 XP_016866037.1 16 - 17 2265 115
XM_017010549.1 XP_016866038.1 16 - 17 2262 115
XM_017010556.1 XP_016866045.1 3 - 4 3327 115
GenBank mRNA AB051866.1 - 24 - 25 3887 115
AF263305.1 - 12 - 13 1353 115
AF263306.1 - 11 - 12 1275 115
AK294868.1 - 19 - 20 2403 115
AK295177.1 - 16 - 17 2164 115
AK302571.1 - 9 - 10 884 115
AY190519.1 - 28 - 29 4131 115

View Full Product Details