Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000051.3 | NP_000042.3 | 36 - 37 | 5883 | 162 |
XM_005271561.4 | XP_005271618.2 | 37 - 38 | 5944 | 162 | |
XM_005271562.4 | XP_005271619.2 | 36 - 37 | 6017 | 162 | |
XM_006718843.3 | XP_006718906.1 | 36 - 37 | 5650 | 162 | |
XM_006718845.1 | XP_006718908.1 | 11 - 12 | 1523 | 162 | |
XM_011542840.2 | XP_011541142.1 | 37 - 38 | 5738 | 162 | |
XM_011542842.2 | XP_011541144.1 | 35 - 36 | 5692 | 162 | |
XM_011542843.2 | XP_011541145.1 | 36 - 37 | 6231 | 162 | |
XM_011542844.2 | XP_011541146.1 | 36 - 37 | 5833 | 162 | |
XM_011542845.1 | XP_011541147.1 | 28 - 29 | 4346 | 162 | |
XM_017017789.1 | XP_016873278.1 | 37 - 38 | 6441 | 162 | |
XM_017017790.1 | XP_016873279.1 | 36 - 37 | 5838 | 162 | |
XM_017017791.1 | XP_016873280.1 | 36 - 37 | 6231 | 162 | |
GenBank mRNA | BC137169.1 | - | 10 - 11 | 1521 | 162 |
CR749436.1 | - | - | 902 | 162 | |
U26455.1 | - | 10 - 11 | 1713 | 162 | |
U33841.1 | - | 37 - 38 | 5687 | 162 |