Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000051.3 | NP_000042.3 | 54 - 55 | 8397 | 142 |
XM_005271561.4 | XP_005271618.2 | 55 - 56 | 8458 | 142 | |
XM_005271562.4 | XP_005271619.2 | 54 - 55 | 8531 | 142 | |
XM_006718843.3 | XP_006718906.1 | 54 - 55 | 8164 | 142 | |
XM_006718845.1 | XP_006718908.1 | 29 - 30 | 4037 | 142 | |
XM_011542840.2 | XP_011541142.1 | 55 - 56 | 8252 | 142 | |
XM_011542842.2 | XP_011541144.1 | 53 - 54 | 8206 | 142 | |
XM_011542843.2 | XP_011541145.1 | 54 - 55 | 8745 | 142 | |
XM_011542844.2 | XP_011541146.1 | 54 - 55 | 8347 | 142 | |
XM_011542845.1 | XP_011541147.1 | 46 - 47 | 6860 | 142 | |
XM_017017789.1 | XP_016873278.1 | 55 - 56 | 8955 | 142 | |
XM_017017790.1 | XP_016873279.1 | 54 - 55 | 8352 | 142 | |
GenBank mRNA | AK299843.1 | - | 16 - 17 | 2228 | 142 |
BC137169.1 | - | 28 - 29 | 4035 | 142 | |
U26455.1 | - | 28 - 29 | 4227 | 142 | |
U33841.1 | - | 55 - 56 | 8201 | 142 |