Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000051.3 | NP_000042.3 | 12 - 13 | 2287 | 141 |
XM_005271561.4 | XP_005271618.2 | 13 - 14 | 2348 | 141 | |
XM_005271562.4 | XP_005271619.2 | 12 - 13 | 2421 | 141 | |
XM_006718843.3 | XP_006718906.1 | 12 - 13 | 2054 | 141 | |
XM_011542840.2 | XP_011541142.1 | 13 - 14 | 2142 | 141 | |
XM_011542842.2 | XP_011541144.1 | 11 - 12 | 2096 | 141 | |
XM_011542843.2 | XP_011541145.1 | 12 - 13 | 2635 | 141 | |
XM_011542844.2 | XP_011541146.1 | 12 - 13 | 2237 | 141 | |
XM_011542845.1 | XP_011541147.1 | 4 - 5 | 750 | 141 | |
XM_017017789.1 | XP_016873278.1 | 13 - 14 | 2845 | 141 | |
XM_017017790.1 | XP_016873279.1 | 12 - 13 | 2242 | 141 | |
XM_017017791.1 | XP_016873280.1 | 12 - 13 | 2635 | 141 | |
XM_017017792.1 | XP_016873281.1 | 12 - 13 | 2635 | 141 | |
GenBank mRNA | U33841.1 | - | 13 - 14 | 2091 | 141 |
X91196.1 | - | 12 - 13 | 2635 | 141 |