Assay Design



Gene Transcripts

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.11: 123358299 - 123627771 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286563.1 NP_001273492.1 15 - 16 2128 67
NM_001286564.1 NP_001273493.1 14 - 15 1837 67
NM_020716.2 NP_065767.1 14 - 15 2107 67
XM_005271619.4 XP_005271676.1 14 - 15 2107 67
XM_005271621.4 XP_005271678.1 15 - 16 6494 67
XM_005271623.4 XP_005271680.1 14 - 15 1872 67
XM_005271624.4 XP_005271681.2 14 - 15 1787 67
XM_006718892.3 XP_006718955.1 15 - 16 2024 67
XM_011542926.2 XP_011541228.1 15 - 16 2021 67
XM_011542929.2 XP_011541231.1 14 - 15 1997 67
XM_011542930.2 XP_011541232.1 14 - 15 2638 67
XM_011542931.2 XP_011541233.1 14 - 15 2552 67
XM_011542933.1 XP_011541235.1 12 - 13 2249 67
XM_017018034.1 XP_016873523.1 16 - 17 2092 67
XM_017018035.1 XP_016873524.1 16 - 17 2089 67
XM_017018036.1 XP_016873525.1 15 - 16 2559 67
XM_017018037.1 XP_016873526.1 16 - 17 2858 67
XM_017018038.1 XP_016873527.1 16 - 17 2092 67
XM_017018039.1 XP_016873528.1 15 - 16 2150 67
XM_017018040.1 XP_016873529.1 14 - 15 1946 67
XM_017018041.1 XP_016873530.1 15 - 16 2150 67
XM_017018042.1 XP_016873531.1 15 - 16 2150 67
XM_017018043.1 XP_016873532.1 14 - 15 1783 67
XM_017018044.1 XP_016873533.1 14 - 15 2107 67
XM_017018045.1 XP_016873534.1 15 - 16 6491 67
XM_017018046.1 XP_016873535.1 15 - 16 2596 67
XM_017018047.1 XP_016873536.1 15 - 16 2593 67
XM_017018048.1 XP_016873537.1 14 - 15 2549 67
XM_017018049.1 XP_016873538.1 15 - 16 2590 67
XM_017018050.1 XP_016873539.1 14 - 15 1866 67
XM_017018051.1 XP_016873540.1 12 - 13 1470 67
GenBank mRNA AB033027.1 - 14 - 15 1770 67
AK127457.1 - 15 - 16 2128 67
AK297606.1 - 14 - 15 1837 67
AY358924.1 - 4 - 5 1360 67
BC105741.1 - 14 - 15 1921 67
BC105931.1 - 14 - 15 1921 67
BC105932.1 - 14 - 15 1921 67
BC107480.1 - 14 - 15 1921 67

View Full Product Details