Assay Design



Gene Transcripts

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.11: 123358299 - 123627771 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286563.1 NP_001273492.1 16 - 17 2328 69
NM_001286564.1 NP_001273493.1 15 - 16 2037 69
NM_020716.2 NP_065767.1 15 - 16 2307 69
XM_005271619.4 XP_005271676.1 15 - 16 2307 69
XM_005271621.4 XP_005271678.1 16 - 17 6694 69
XM_005271623.4 XP_005271680.1 15 - 16 2072 69
XM_005271624.4 XP_005271681.2 15 - 16 1987 69
XM_006718892.3 XP_006718955.1 16 - 17 2224 69
XM_011542926.2 XP_011541228.1 16 - 17 2221 69
XM_011542929.2 XP_011541231.1 15 - 16 2197 69
XM_011542930.2 XP_011541232.1 15 - 16 2838 69
XM_011542931.2 XP_011541233.1 15 - 16 2752 69
XM_011542933.1 XP_011541235.1 13 - 14 2449 69
XM_017018034.1 XP_016873523.1 17 - 18 2292 69
XM_017018035.1 XP_016873524.1 17 - 18 2289 69
XM_017018036.1 XP_016873525.1 16 - 17 2759 69
XM_017018037.1 XP_016873526.1 17 - 18 3058 69
XM_017018038.1 XP_016873527.1 17 - 18 2292 69
XM_017018039.1 XP_016873528.1 16 - 17 2350 69
XM_017018040.1 XP_016873529.1 15 - 16 2146 69
XM_017018041.1 XP_016873530.1 16 - 17 2350 69
XM_017018042.1 XP_016873531.1 16 - 17 2350 69
XM_017018043.1 XP_016873532.1 15 - 16 1983 69
XM_017018044.1 XP_016873533.1 15 - 16 2307 69
XM_017018045.1 XP_016873534.1 16 - 17 6691 69
XM_017018046.1 XP_016873535.1 16 - 17 2796 69
XM_017018047.1 XP_016873536.1 16 - 17 2793 69
XM_017018048.1 XP_016873537.1 15 - 16 2749 69
XM_017018049.1 XP_016873538.1 16 - 17 2790 69
XM_017018050.1 XP_016873539.1 15 - 16 2066 69
XM_017018051.1 XP_016873540.1 13 - 14 1670 69
GenBank mRNA AB033027.1 - 15 - 16 1970 69
AK127457.1 - 16 - 17 2328 69
AK297606.1 - 15 - 16 2037 69
AY358924.1 - 5 - 6 1560 69
BC105741.1 - 15 - 16 2121 69
BC105931.1 - 15 - 16 2121 69
BC105932.1 - 15 - 16 2121 69
BC107480.1 - 15 - 16 2121 69

View Full Product Details