Assay Design



Gene Transcripts

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.11: 123358299 - 123627771 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286564.1 NP_001273493.1 2 - 3 473 158
NM_020716.2 NP_065767.1 2 - 3 743 158
XM_005271619.4 XP_005271676.1 2 - 3 743 158
XM_005271621.4 XP_005271678.1 3 - 4 5130 158
XM_005271623.4 XP_005271680.1 2 - 3 508 158
XM_005271624.4 XP_005271681.2 2 - 3 423 158
XM_006718892.3 XP_006718955.1 3 - 4 660 158
XM_011542926.2 XP_011541228.1 3 - 4 657 158
XM_011542929.2 XP_011541231.1 2 - 3 633 158
XM_011542930.2 XP_011541232.1 2 - 3 1274 158
XM_011542931.2 XP_011541233.1 2 - 3 1188 158
XM_017018034.1 XP_016873523.1 4 - 5 728 158
XM_017018035.1 XP_016873524.1 4 - 5 725 158
XM_017018036.1 XP_016873525.1 3 - 4 1195 158
XM_017018037.1 XP_016873526.1 4 - 5 1494 158
XM_017018038.1 XP_016873527.1 4 - 5 728 158
XM_017018039.1 XP_016873528.1 3 - 4 786 158
XM_017018040.1 XP_016873529.1 2 - 3 582 158
XM_017018041.1 XP_016873530.1 3 - 4 786 158
XM_017018042.1 XP_016873531.1 3 - 4 786 158
XM_017018043.1 XP_016873532.1 2 - 3 419 158
XM_017018044.1 XP_016873533.1 2 - 3 743 158
XM_017018045.1 XP_016873534.1 3 - 4 5127 158
XM_017018046.1 XP_016873535.1 3 - 4 1232 158
XM_017018047.1 XP_016873536.1 3 - 4 1229 158
XM_017018048.1 XP_016873537.1 2 - 3 1185 158
XM_017018049.1 XP_016873538.1 3 - 4 1226 158
XM_017018050.1 XP_016873539.1 2 - 3 502 158
GenBank mRNA AB033027.1 - 2 - 3 406 158
AK297606.1 - 2 - 3 473 158
AK307795.1 - 2 - 3 583 158
BC105741.1 - 2 - 3 557 158
BC105931.1 - 2 - 3 557 158
BC105932.1 - 2 - 3 557 158
BC107480.1 - 2 - 3 557 158

View Full Product Details