Assay Design



Gene Transcripts

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.11: 123358299 - 123627771 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286563.1 NP_001273492.1 5 - 6 872 117
NM_001286564.1 NP_001273493.1 4 - 5 581 117
NM_020716.2 NP_065767.1 4 - 5 851 117
XM_005271619.4 XP_005271676.1 4 - 5 851 117
XM_005271621.4 XP_005271678.1 5 - 6 5238 117
XM_005271623.4 XP_005271680.1 4 - 5 616 117
XM_005271624.4 XP_005271681.2 4 - 5 531 117
XM_006718892.3 XP_006718955.1 5 - 6 768 117
XM_011542926.2 XP_011541228.1 5 - 6 765 117
XM_011542929.2 XP_011541231.1 4 - 5 741 117
XM_011542930.2 XP_011541232.1 4 - 5 1382 117
XM_011542931.2 XP_011541233.1 4 - 5 1296 117
XM_011542933.1 XP_011541235.1 2 - 3 993 117
XM_017018034.1 XP_016873523.1 6 - 7 836 117
XM_017018035.1 XP_016873524.1 6 - 7 833 117
XM_017018036.1 XP_016873525.1 5 - 6 1303 117
XM_017018037.1 XP_016873526.1 6 - 7 1602 117
XM_017018038.1 XP_016873527.1 6 - 7 836 117
XM_017018039.1 XP_016873528.1 5 - 6 894 117
XM_017018040.1 XP_016873529.1 4 - 5 690 117
XM_017018041.1 XP_016873530.1 5 - 6 894 117
XM_017018042.1 XP_016873531.1 5 - 6 894 117
XM_017018043.1 XP_016873532.1 4 - 5 527 117
XM_017018044.1 XP_016873533.1 4 - 5 851 117
XM_017018045.1 XP_016873534.1 5 - 6 5235 117
XM_017018046.1 XP_016873535.1 5 - 6 1340 117
XM_017018047.1 XP_016873536.1 5 - 6 1337 117
XM_017018048.1 XP_016873537.1 4 - 5 1293 117
XM_017018049.1 XP_016873538.1 5 - 6 1334 117
XM_017018050.1 XP_016873539.1 4 - 5 610 117
XM_017018051.1 XP_016873540.1 2 - 3 214 117
GenBank mRNA AB033027.1 - 4 - 5 514 117
AK127457.1 - 5 - 6 872 117
AK297606.1 - 4 - 5 581 117
AK307795.1 - 4 - 5 691 117
BC105741.1 - 4 - 5 665 117
BC105931.1 - 4 - 5 665 117
BC105932.1 - 4 - 5 665 117
BC107480.1 - 4 - 5 665 117

View Full Product Details