Assay Design



Gene Transcripts

Gene Symbol
CCDC84
Entrez Gene ID
338657
Gene Name
coiled-coil domain containing 84
Gene Aliases
DLNB14
Chromosome Location
Chr.11: 118998067 - 119015793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198489.2 NP_940891.1 2 - 3 347 107
NR_104049.1 2 - 3 347 107
NR_104050.1 2 - 3 347 107
NR_104051.1 2 - 3 347 107
XM_011542799.1 XP_011541101.1 1 - 2 162 107
XM_017017652.1 XP_016873141.1 2 - 3 413 107
GenBank mRNA AB094093.1 - 2 - 3 338 107
BC126469.1 - 2 - 3 312 107
BC126471.1 - 2 - 3 312 107
BC144208.1 - 2 - 3 312 107
BC144209.1 - 2 - 3 312 107
BC144210.1 - 2 - 3 312 107
HY073607.1 - 2 - 3 349 107

View Full Product Details