Assay Design



Gene Transcripts

Gene Symbol
CCDC84
Entrez Gene ID
338657
Gene Name
coiled-coil domain containing 84
Gene Aliases
DLNB14
Chromosome Location
Chr.11: 118998067 - 119015793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198489.2 NP_940891.1 6 - 7 639 68
NR_104049.1 6 - 7 667 68
NR_104050.1 6 - 7 667 68
NR_104051.1 7 - 8 880 68
XM_011542799.1 XP_011541101.1 6 - 7 667 68
XM_011542800.2 XP_011541102.1 4 - 5 347 68
XM_017017652.1 XP_016873141.1 6 - 7 3421 68
GenBank mRNA AB094093.1 - 6 - 7 630 68
BC126469.1 - 6 - 7 604 68
BC126471.1 - 6 - 7 604 68
BC144208.1 - 7 - 8 845 68
BC144209.1 - 6 - 7 632 68
BC144210.1 - 6 - 7 632 68

View Full Product Details