Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024383.1 | NP_001019554.1 | 5 - 6 | 841 | 70 |
NM_014903.5 | NP_055718.4 | 5 - 6 | 841 | 70 | |
XM_005269215.3 | XP_005269272.1 | 5 - 6 | 4842 | 70 | |
XM_011538940.2 | XP_011537242.1 | 5 - 6 | 4843 | 70 | |
XM_011538941.2 | XP_011537243.1 | 5 - 6 | 4842 | 70 | |
XM_011538944.2 | XP_011537246.1 | 5 - 6 | 4840 | 70 | |
XM_017020164.1 | XP_016875653.1 | 5 - 6 | 4844 | 70 | |
XM_017020165.1 | XP_016875654.1 | 5 - 6 | 4844 | 70 | |
XM_017020166.1 | XP_016875655.1 | 5 - 6 | 830 | 70 | |
XM_017020168.1 | XP_016875657.1 | 5 - 6 | 4843 | 70 | |
XM_017020169.1 | XP_016875658.1 | 5 - 6 | 4842 | 70 | |
XM_017020170.1 | XP_016875659.1 | 5 - 6 | 4839 | 70 | |
GenBank mRNA | AF397731.1 | - | 4 - 5 | 425 | 70 |
AJ488201.1 | - | 5 - 6 | 713 | 70 | |
BC017667.2 | - | 5 - 6 | 1218 | 70 |