Assay Design



Gene Transcripts

Gene Symbol
RIN3
Entrez Gene ID
79890
Gene Name
Ras and Rab interactor 3
Gene Aliases
-
Chromosome Location
Chr.14: 92513773 - 92688994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011537165.2 XP_011535467.1 3 - 4 522 101

View Full Product Details