Assay Design



Gene Transcripts

Gene Symbol
SLAIN1
Entrez Gene ID
122060
Gene Name
SLAIN motif family member 1
Gene Aliases
C13orf32
Chromosome Location
Chr.13: 77697854 - 77764242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040153.3 NP_001035243.2 4 - 5 805 67
NM_001242868.1 NP_001229797.1 4 - 5 1314 67
NM_001242871.1 NP_001229800.1 3 - 4 853 67
NM_144595.3 NP_653196.1 3 - 4 769 67
XM_011534929.2 XP_011533231.1 5 - 6 1194 67
XM_011534930.1 XP_011533232.1 4 - 5 707 67
GenBank mRNA AK054608.1 - 3 - 4 549 67
AK294201.1 - 4 - 5 805 67
AK295423.1 - 3 - 4 629 67
AL834332.1 - 3 - 4 562 67
BC045177.1 - 3 - 4 578 67
BC111497.1 - 3 - 4 560 67

View Full Product Details