Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201427.1 | NP_001188356.1 | 9 - 10 | 1180 | 74 |
NM_015345.3 | NP_056160.2 | 9 - 10 | 1244 | 74 | |
XM_006715039.3 | XP_006715102.3 | 9 - 10 | 1825 | 74 | |
XM_006715040.3 | XP_006715103.3 | 9 - 10 | 1824 | 74 | |
XM_006715042.2 | XP_006715105.1 | 9 - 10 | 3820 | 74 | |
XM_006715043.2 | XP_006715106.1 | 9 - 10 | 1197 | 74 | |
XM_006715045.3 | XP_006715108.1 | 9 - 10 | 2412 | 74 | |
XM_006715046.3 | XP_006715109.1 | 9 - 10 | 1590 | 74 | |
XM_017010630.1 | XP_016866119.1 | 9 - 10 | 2436 | 74 | |
GenBank mRNA | AB002379.2 | - | 9 - 10 | 1180 | 74 |
AK310941.1 | - | 9 - 10 | 1187 | 74 | |
AL833083.2 | - | 9 - 10 | 1224 | 74 |